missing translation for 'onlineSavingsMsg'
Learn More

Sodium Formate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02004093
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02004093 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02004093 Supplier Avantor Supplier No. 370005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 141-53-7
Physical Form Crystals
Quantity 2.5 kg
Molecular Formula CHNaO2
Synonym sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
SMILES C(=O)[O-].[Na+]
IUPAC Name sodium;formate
Molecular Weight (g/mol) 68.007
PubChem CID 2723810
ChEBI CHEBI:62965
Formula Weight 68.01
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.92kg
Chemical Name or Material Sodium Formate
Show More Show Less