missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ethoxide 95.0+%, TCI America™

Catalog No. S0547100G
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0547100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0547100G Supplier TCI America Supplier No. S0547100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium ethoxide suitable for chemical synthesis and laboratory research applications.

Chemical Identifiers

CAS 141-52-6
Molecular Formula C2H5NaO
Molecular Weight (g/mol) 68.051
MDL Number MFCD00012417
InChI Key QDRKDTQENPPHOJ-UHFFFAOYSA-N
Synonym sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1
PubChem CID 2723922
ChEBI CHEBI:52096
IUPAC Name sodium;ethanolate
SMILES CC[O-].[Na+]

Specifications

Melting Point 260°C
Physical Form Crystalline Powder
Quantity 100 g
UN Number 3206
Formula Weight 68.05
Percent Purity ≥95.0% (T)
Chemical Name or Material Sodium Ethoxide
EINECSNumber (2)-0204
TSCA Yes