missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ethoxide (ca. 20% in Ethanol), TCI America™

Catalog No. S0548500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0548500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0548500ML Supplier TCI America Supplier No. S0548500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium ethoxide solution for chemical synthesis.

Chemical Identifiers

CAS 141-52-6
Molecular Formula C2H5NaO
Molecular Weight (g/mol) 68.051
MDL Number MFCD00012417
InChI Key QDRKDTQENPPHOJ-UHFFFAOYSA-N
Synonym sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1
PubChem CID 2723922
ChEBI CHEBI:52096
IUPAC Name sodium;ethanolate
SMILES CC[O-].[Na+]

Specifications

Color Yellow
Physical Form Liquid
Quantity 500 mL
UN Number 3274
Formula Weight 68.05
Chemical Name or Material Sodium Ethoxide (ca. 20% in Ethanol)
EINECSNumber (2)-0204
TSCA Yes