missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecylsulfate, 10% Solution, Ultrapure
CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
$129.53 - $332.16
Chemical Identifiers
| CAS | 151-21-3 |
|---|---|
| Molecular Formula | C12H25NaO4S |
| Molecular Weight (g/mol) | 288.38 |
| MDL Number | MFCD00036175 |
| InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
| PubChem CID | 3423265 |
| ChEBI | CHEBI:8984 |
| SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AAJ77504AE
|
N/A | 100 mL | Nalgene bottle |
Each for $129.53
|
|
||||
|
AAJ77504AP
|
N/A | 500 mL | Nalgene bottle |
Each for $298.95
|
|
||||
|
AAJ77504K2
|
N/A | 1 L | Nalgene bottle |
Each for $332.16
|
|
||||
Chemical Identifiers
| 151-21-3 | |
| 288.38 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| CHEBI:8984 |
| C12H25NaO4S | |
| MFCD00036175 | |
| 3423265 | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Specifications
| 151-21-3 | |
| 10% Solution | |
| MFCD00036175 | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 288.38 | |
| CHEBI:8984 |
| Solution | |
| C12H25NaO4S | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| sodium dodecyl sulfate | |
| 3423265 | |
| Sodium Dodecylsulfate |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
H315-H318
EINECSNumber : 205-788-1
RTECSNumber : WT1050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only