missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecylsulfate, 10% Solution, Ultrapure

Catalog No. AAJ77504K2
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ77504K2 1 L Nalgene bottle
AAJ77504AE 100 mL Nalgene bottle
AAJ77504AP 500 mL Nalgene bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ77504K2 Supplier Thermo Fisher Scientific Supplier No. J77504K2

missing translation for 'validMainframeMsg'

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Physical Form Solution
Concentration 10% Solution
Quantity 1 L
Packaging Nalgene bottle
IUPAC Name sodium;dodecyl sulfate
Chemical Name or Material Sodium Dodecylsulfate
Hazard Category H315-H318
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
EINECSNumber 205-788-1
RTECSNumber WT1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only