missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecylsulfate, 10% Solution, Ultrapure
CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
Supplier: Thermo Fisher Scientific J77504AP
| Quantity | 500 mL |
|---|---|
| Packaging | Nalgene bottle |
Chemical Identifiers
| 151-21-3 | |
| 288.38 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| CHEBI:8984 |
| C12H25NaO4S | |
| MFCD00036175 | |
| 3423265 | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Specifications
| Sodium Dodecylsulfate | |
| Solution | |
| 500 mL | |
| C12H25NaO4S | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| sodium dodecyl sulfate | |
| 3423265 |
| 151-21-3 | |
| 10% Solution | |
| Nalgene bottle | |
| MFCD00036175 | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 288.38 | |
| CHEBI:8984 |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
H315-H318
EINECSNumber : 205-788-1
RTECSNumber : WT1050000
TSCA : Yes
Recommended Storage : Ambient temperatures