missing translation for 'onlineSavingsMsg'
Learn More

Sodium dodecyl sulfate, >99%, MP Biomedicals™

Catalog No. ICN10291810
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10291810 10 g
ICN10291880 100 g
ICN10291890 500 g
ICN10291891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10291810 Supplier MP Biomedicals Inc Supplier No. 0210291810
Only null left

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Color White
Density >1.1 at 20°C
Quantity 10 g
IUPAC Name sodium dodecyl sulfate
Formula Weight 288.38
Percent Purity >99%
Physical Form Granular Powder
Chemical Name or Material Sodium dodecyl sulfate