missing translation for 'onlineSavingsMsg'
Learn More

Sodium dodecyl sulfate, 99.5%, MP Biomedicals™

$95.01 - $751.83

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium lauryl sulfate, SDS
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19483125
View Documents
MP Biomedicals Inc
0219483125
25 g
Each for $95.01
Only null left
 
ICN19483180
View Documents
MP Biomedicals Inc
0219483180
100 g
Each for $140.51
Only null left
 
ICN19483190
missing translation for 'viewDocuments'
MP Biomedicals Inc
0219483190
500 g
Each for $553.61
Only null left
 
ICN19483191
missing translation for 'viewDocuments'
MP Biomedicals Inc
0219483191
1 kg
Each for $751.83
Only null left
 
Description

Description

Specifications

Chemical Identifiers

151-21-3
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
3423265
sodium dodecyl sulfate
C12H25NaO4S
MFCD00036175
Sodium lauryl sulfate, SDS
CHEBI:8984
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

White
Granular Powder
C12H25NaO4S
Sodium lauryl sulfate, SDS
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
CHEBI:8984
99.5%
151-21-3
25 g
MFCD00036175
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
3423265
288.38
Sodium dodecyl sulfate
Videos
SDS
Documents

Documents