missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium dodecyl sulfate, 99.5%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0219483190
Specifications
| Sodium dodecyl sulfate | |
| 151-21-3 | |
| 500 g | |
| MFCD00036175 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| sodium dodecyl sulfate | |
| 3423265 | |
| 288.38 |
| White | |
| Granular Powder | |
| C12H25NaO4S | |
| Sodium lauryl sulfate, SDS | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 288.38 | |
| CHEBI:8984 | |
| 99.5% |
Chemical Identifiers
| 151-21-3 | |
| 288.38 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| 3423265 | |
| sodium dodecyl sulfate |
| C12H25NaO4S | |
| MFCD00036175 | |
| Sodium lauryl sulfate, SDS | |
| CHEBI:8984 | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |