missing translation for 'onlineSavingsMsg'
Learn More

Sodium dodecyl sulfate, 99.5%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219483180

 View more versions of this product

Catalog No. ICN19483180


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium dodecyl sulfate
151-21-3
100 g
MFCD00036175
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
3423265
288.38
White
Granular Powder
C12H25NaO4S
Sodium lauryl sulfate, SDS
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
CHEBI:8984
99.5%

Chemical Identifiers

151-21-3
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
3423265
sodium dodecyl sulfate
C12H25NaO4S
MFCD00036175
Sodium lauryl sulfate, SDS
CHEBI:8984
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions