missing translation for 'onlineSavingsMsg'
Learn More

SDS Solution, 10% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™

Supplier:  Fisher BioReagents BP2436 1

Quantity 1 L
Packaging Nalgene Poly Bottle

 View more versions of this product

Catalog No. BP24361



Get 1 Extra Chemical When You Buy 2
View all promotions
Only null left

Description

Description

  • DNase, RNase, and protease-free premixed solution containing 10% SDS for protein extraction and DNA extraction procedures
  • Reduce preparation time by using this premade SDS solution
  • SDS is a commonly used detergent in protein purification, polyacrylamide gel electrophoresis (PAGE), and other molecular biology applications
  • SDS is also used to disrupt cell membranes and dissociate nucleic acid-protein complexes in DNA extraction procedures
  • Intended for research use only; not for human or animal diagnostic purposes
Specifications

Chemical Identifiers

151-21-3 , 7732-18-5
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
CHEBI:8984
C12H25NaO4S
MFCD00036175
3423265
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Sodium Dodecyl Sulfate
Colorless
151-21-3 , 7732-18-5
100°C
9.5 to 10.5%
Pass Test
C12H25NaO4S
MFCD00036175
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium;dodecyl sulfate
3423265
288.38
0.4 max. (0.1% solution) at 230nm, 0.1 max. (0.1% solution) at 280nm
10% Solution
Liquid
1.01g/cm³
1 L
Nalgene Poly Bottle
CH3(CH2)10CH2OSO3Na
15, 8768
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
CHEBI:8984
Electrophoresis
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS
Documents

Documents

Promotions

Promotions