missing translation for 'onlineSavingsMsg'
Learn More

SDS Solution, 10% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™

Catalog No. p-28628
Change view
Click to view available options
Quantity:
200 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2436200 200 mL Poly Bottle
BP24361 1 L Nalgene Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP2436200 Supplier Fisher BioReagents Supplier No. BP2436 200
Only null left
  • DNase, RNase, and protease-free premixed solution containing 10% SDS for protein extraction and DNA extraction procedures
  • Reduce preparation time by using this premade SDS solution
  • SDS is a commonly used detergent in protein purification, polyacrylamide gel electrophoresis (PAGE), and other molecular biology applications
  • SDS is also used to disrupt cell membranes and dissociate nucleic acid-protein complexes in DNA extraction procedures
  • Intended for research use only; not for human or animal diagnostic purposes

Chemical Identifiers

CAS 151-21-3 , 7732-18-5
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Absorbance 0.4 max. (0.1% solution) at 230nm, 0.1 max. (0.1% solution) at 280nm
Color Colorless
Physical Form Liquid
Boiling Point 100°C
Density 1.01g/cm³
Concentration 9.5 to 10.5%
Quantity 200 mL
Identification Pass Test
Packaging Poly Bottle
Linear Formula CH3(CH2)10CH2OSO3Na
Merck Index 15, 8768
IUPAC Name sodium dodecyl sulfate
Formula Weight 288.38
Grade Electrophoresis
Chemical Name or Material Sodium Dodecyl Sulfate, 10% Solution
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT