missing translation for 'onlineSavingsMsg'
Learn More

Sodium DL-3-Hydroxybutyrate 99.0+%, TCI America™

Catalog No. H023125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H023125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H023125G Supplier TCI America Supplier No. H023125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 150-83-4
Molecular Formula C4H7NaO3
Molecular Weight (g/mol) 126.087
MDL Number MFCD00016716
InChI Key NBPUSGBJDWCHKC-UHFFFAOYSA-M
Synonym sodium 3-hydroxybutyrate, sodium 3-hydroxybutanoate, dl-3-hydroxybutyric acid sodium salt, sodium beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, monosodium salt, butanoic acid,3-hydroxy-, sodium salt 1:1, dl-3-hydroxybutyric acid, sodium salt, 3-hydroxybutyric acid sodium salt, sodium 3-oxidanylbutanoate, dl-beta-hydroxybutyric acid sodium salt
PubChem CID 23676771
IUPAC Name sodium;3-hydroxybutanoate
SMILES CC(CC(=O)[O-])O.[Na+]

Specifications

Melting Point 170°C
Color White
Quantity 25 g
Formula Weight 126.09
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium DL-3-Hydroxybutyrate