missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dithionite (Sodium Hydrosulfite), Reagents

Catalog No. 01337024
Change view
Click to view available options
Quantity:
500 g
2.5 kg
100 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337024 100 kg
01337023 2.5 kg
01337022 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337024 Supplier Labchem Inc Supplier No. C2316300100KG

Sodium Dithionite (Sodium Hydrosulfite), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7775-14-6
Molecular Formula Na2O4S2
Molecular Weight (g/mol) 174.10
InChI Key JVBXVOWTABLYPX-UHFFFAOYSA-L
Synonym Sodium Hydrosulfite
IUPAC Name disodium sulfinatosulfinate
SMILES [Na+].[Na+].[O-]S(=O)S([O-])=O

Specifications

Boiling Point >80°C
Melting Point 52°C
Color White to Off-White
pH 8 to 10.5
Physical Form Solid
Quantity 100 kg
Grade Pure
Packaging Steel Drum
Specific Gravity 2.19
Chemical Name or Material Sodium Dithionite (Sodium Hydrosulfite)