missing translation for 'onlineSavingsMsg'
Learn More

Sodium Diphenylamine Sulfonate Solution, Reagents

Catalog No. 01334004
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334004 1 L
01334003 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334004 Supplier Labchem Inc Supplier No. CS0060001A

missing translation for 'validMainframeMsg'

Sodium Diphenylamine Sulfonate Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Diphenylamine Sulfonate Solution
Packaging Poly Bottle
CAS 7732-18-5, 7664-93-9, 7664-38-2
Specific Gravity 1.0121
Physical Form Liquid
Color Green
Boiling Point 100°C
Synonym Sodium Diphenylaminesulfonic Acid
Solubility Completely miscible
Quantity 1 L
Grade Reagent
Show More Show Less