Learn More
Sodium dihydrogen phosphate monohydrate, ACS, 98.0-102.0%
Description
Sodium phosphate monobasic monohydrate is used as sequestrant, emulsifier, mordant in dyeing, reagent and buffer in foods and analytical chemistry. It is applied as a fireproofing agent and for weighting silk in tanning. It is employed in manufacturing of enamels, ceramics, detergents, boiler compounds, in soldering and brazing instead of borax.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 100°C-H2O |
| Color | White |
| Physical Form | Crystalline |
| Quantity | 5 kg |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Appearance | White crystals |
| Assay Percent Range | 98.0-102.0% |
| Solubility Information | Highly soluble in water. Insoluble in ethanol,ether and chloroform. |
| IUPAC Name | sodium phosphoric acid hydrate |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. |
| EINECSNumber | 231-449-2 |
| RTECSNumber | WA1900000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only