missing translation for 'onlineSavingsMsg'
Learn More

Sodium diethyldithiocarbamate trihydrate, 98%

Catalog No. AAA1589836
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1589836 500 g
AAA1589822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1589836 Supplier Thermo Scientific Chemicals Supplier No. A1589836

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20624-25-3 | C5H16NNaO3S2 | 225.297 g/mol

Sodium diethyldithiocarbamate trihydrate is a spin trapping reagent used in conjunction with ferrous ion to detect nitric oxide in brain, kidney, liver and other tissues. It is used as sensitive reagent for the determination of copper, rubber vulcanization accelerator and finds application in the determination of other metals such as zinc, cobalt, platinum and palladium. Furthermore, it is an inhibitor of superoxide dismutase, ascorbate oxidase and other enzymes as well as inhibits the induction of macrophage nitric oxide synthase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium diethyldithiocarbamate trihydrate is a spin trapping reagent used in conjunction with ferrous ion to detect nitric oxide in brain, kidney, liver and other tissues. It is used as sensitive reagent for the determination of copper, rubber vulcanization accelerator and finds application in the determination of other metals such as zinc, cobalt, platinum and palladium. Furthermore, it is an inhibitor of superoxide dismutase, ascorbate oxidase and other enzymes as well as inhibits the induction of macrophage nitric oxide synthase.

Solubility
Soluble in water, ethanol, methanol and acetone. Insoluble in ether.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 20624-25-3
Molecular Formula C5H16NNaO3S2
Molecular Weight (g/mol) 225.297
MDL Number MFCD00150617
InChI Key WWGXHTXOZKVJDN-UHFFFAOYSA-M
Synonym sodium diethyldithiocarbamate trihydrate, dithiocarb trihydrate, cupral, diethyldithiocarbamic acid sodium salt trihydrate, ditiocarb sodium trihydrate, unii-04574exf2c, diethyldithiocarbamate sodium trihydrate, carbamic acid, diethyldithio-, sodium salt, trihydrate, diethyldithiocarbamate sodium salt trihydrate, diethyldithiokarbaman sodny trihydrat czech
PubChem CID 517546
IUPAC Name sodium;N,N-diethylcarbamodithioate;trihydrate
SMILES CCN(CC)C(=S)[S-].O.O.O.[Na+]

Specifications

Melting Point 94°C to 102°C
Quantity 500 g
Beilstein 3920507
Sensitivity Hygroscopic
Merck Index 14,3378
Solubility Information Soluble in water,ethanol,methanol and acetone. Insoluble in ether.
Formula Weight 225.31 (171.28 Anhydrous)
Percent Purity 98%
Chemical Name or Material Sodium diethyldithiocarbamate trihydrate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 205-710-6
RTECSNumber EZ6550000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only