missing translation for 'onlineSavingsMsg'
Learn More

Sodium Diethyldithiocarbamate, 0.1% (w/v), Ricca Chemical

Catalog No. R72406001A
Change view
Click to view available options
Package: Natural Poly; Size: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
R72406001A Package: Natural Poly; Size: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R72406001A Supplier Ricca Chemical Company Supplier No. R72406001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS/ASTM E 200 Reagent Solution

Specifications

CAS 20624-25-3, 7732-18-5
Assay Percent Range 0.1% (w/v)
Molecular Formula C5H16NNaO3S2
Synonym sodium diethyldithiocarbamate trihydrate, dithiocarb trihydrate, cupral, diethyldithiocarbamic acid sodium salt trihydrate, ditiocarb sodium trihydrate, unii-04574exf2c, diethyldithiocarbamate sodium trihydrate, carbamic acid, diethyldithio-, sodium salt, trihydrate, diethyldithiocarbamate sodium salt trihydrate, diethyldithiokarbaman sodny trihydrat czech
InChI Key WWGXHTXOZKVJDN-UHFFFAOYSA-M
SMILES CCN(CC)C(=S)[S-].O.O.O.[Na+]
IUPAC Name sodium;N,N-diethylcarbamodithioate;trihydrate
Molecular Weight (g/mol) 225.297
PubChem CID 517546
Grade ASTM/ACS Certified Reagent
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 1 L
Chemical Name or Material Sodium Diethyldithiocarbamate, 0.1% (w/v) Aqueous Solution
Show More Show Less