missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dicyanamide 98.0+%, TCI America™

Catalog No. S083825G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S083825G 25 g
S0838100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S083825G Supplier TCI America Supplier No. S083825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1934-75-4
Molecular Formula C2N3Na
Molecular Weight (g/mol) 89.03
MDL Number MFCD00003535
InChI Key IXBPPZBJIFNGJJ-UHFFFAOYSA-N
Synonym sodium dicyanamide, sodium dicyanoazanide, sodium dicyanoamide, unii-i9c96i1mre, cyanamide, cyano-, sodium salt, cyanamide,cyano-,sodium salt, i9c96i1mre, cyanamide, n-cyano-, sodium salt 1:1, dicyanamide, sodium salt, nadca
PubChem CID 4350929
IUPAC Name sodium dicyanoazanide
SMILES [Na+].N#C[N-]C#N

Specifications

Melting Point 260°C
Color White
Quantity 25 g
Formula Weight 89.03
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Dicyanamide