missing translation for 'onlineSavingsMsg'
Learn More

Sodium dicyanamide, 96%

Catalog No. AAB2163436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2163436 500 g
AAB2163414 25 g
AAB2163422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2163436 Supplier Thermo Scientific Chemicals Supplier No. B2163436

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1934-75-4 | C2N3Na | 89.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1934-75-4
Molecular Formula C2N3Na
Molecular Weight (g/mol) 89.03
MDL Number MFCD00003535
InChI Key IXBPPZBJIFNGJJ-UHFFFAOYSA-N
Synonym sodium dicyanamide, sodium dicyanoazanide, sodium dicyanoamide, unii-i9c96i1mre, cyanamide, cyano-, sodium salt, cyanamide,cyano-,sodium salt, i9c96i1mre, cyanamide, n-cyano-, sodium salt 1:1, dicyanamide, sodium salt, nadca
PubChem CID 4350929
IUPAC Name sodium;dicyanoazanide
SMILES [Na+].N#C[N-]C#N

Specifications

Melting Point >300°C
Quantity 500 g
Sensitivity Hygroscopic
Formula Weight 89.03
Percent Purity 96%
Chemical Name or Material Sodium dicyanamide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 217-703-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only