missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dichloroisocyanurate 97.0+%, TCI America™

Catalog No. D100325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D100325G 25 g
D1003500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D100325G Supplier TCI America Supplier No. D100325G

Chemical Identifiers

CAS 2893-78-9
Molecular Formula C3HCl2N3NaO3
Molecular Weight (g/mol) 220.949
MDL Number MFCD00006036
InChI Key UNWRHVZXVVTASG-UHFFFAOYSA-N
Synonym sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt
PubChem CID 86657659
IUPAC Name 1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium
SMILES C1(=O)NC(=O)N(C(=O)N1Cl)Cl.[Na]

Specifications

Color White
Quantity 25 g
UN Number 2465
Formula Weight 219.94
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Dichloroisocyanurate
EINECSNumber (5)-1043
RTECSNumber XZ1900000
TSCA Yes