missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dichloroacetate 99.0+%, TCI America™

Catalog No. D171925G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D171925G 25 g
D171910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D171925G Supplier TCI America Supplier No. D171925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2156-56-1
Molecular Formula C2HCl2NaO2
Molecular Weight (g/mol) 150.92
MDL Number MFCD00070489
InChI Key LUPNKHXLFSSUGS-UHFFFAOYSA-M
Synonym sodium dichloroacetate, sodium 2,2-dichloroacetate, dichloroacetic acid sodium salt, acetic acid, dichloro-, sodium salt, ceresine, dichloroacetate sodium, dichloroctan sodny czech, sodium dichloroacetate usan, ccris 7697, dichloroacetic acid, sodium salt
PubChem CID 517326
IUPAC Name sodium 2,2-dichloroacetate
SMILES [Na+].[O-]C(=O)C(Cl)Cl

Specifications

Melting Point 198°C
Color White
pH 6.0 to 8.5 (50g/L H2O 25°C)
Quantity 25 g
Formula Weight 150.92
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Dichloroacetate
RTECSNumber AG9275000
TSCA No