missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dichloroacetate 97.0+%, TCI America™

Catalog No. D104825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D104825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D104825G Supplier TCI America Supplier No. D104825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2156-56-1
Molecular Formula C2HCl2NaO2
Molecular Weight (g/mol) 150.92
MDL Number MFCD00070489
InChI Key LUPNKHXLFSSUGS-UHFFFAOYSA-M
Synonym sodium dichloroacetate, sodium 2,2-dichloroacetate, dichloroacetic acid sodium salt, acetic acid, dichloro-, sodium salt, ceresine, dichloroacetate sodium, dichloroctan sodny czech, sodium dichloroacetate usan, ccris 7697, dichloroacetic acid, sodium salt
PubChem CID 517326
IUPAC Name sodium 2,2-dichloroacetate
SMILES [Na+].[O-]C(=O)C(Cl)Cl

Specifications

Melting Point 198°C
Color White
pH 4.5 - 8.5
Quantity 25 g
Formula Weight 150.92
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Dichloroacetate
RTECSNumber AG9275000
TSCA No