missing translation for 'onlineSavingsMsg'
Learn More

Sodium diatrizoate, 99%

Catalog No. AC460960050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460960050 5 g
AC460960250 25 g
AC460961000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC460960050 Supplier Thermo Scientific Chemicals Supplier No. 460960050
Only null left

CAS: 737-31-5 | C11H8I3N2NaO4 | 635.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 737-31-5
Molecular Formula C11H8I3N2NaO4
Molecular Weight (g/mol) 635.9
InChI Key ZEYOIOAKZLALAP-UHFFFAOYSA-M
Synonym diatrizoate sodium, sodium diatrizoate, sodium amidotrizoate, hypaque sodium, triombrin, triombrine, hypaque, vascoray, hypaque cysto, hpaque-cysto
PubChem CID 23672589
ChEBI CHEBI:53692
IUPAC Name sodium;3,5-diacetamido-2,4,6-triiodobenzoate
SMILES CC(=O)NC1=C(C(=C(C(=C1I)C(=O)[O-])I)NC(=O)C)I.[Na+]

Specifications

Melting Point 261.0°C to 262.0°C
Quantity 5 g
Solubility Information Solubility in water: 600g/L (20°C).
Formula Weight 635.9
Percent Purity 99%
Infrared Spectrum Authentic
Loss on Drying 7% max.
Chemical Name or Material Sodium diatrizoate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
EINECSNumber 212-004-1

RUO – Research Use Only