missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyanide, 10% (w/v), Ricca Chemical

Catalog No. 252532
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
252532 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 252532 Supplier Ricca Chemical Company Supplier No. 252532
Only null left

Chemical Identifiers

CAS 7732-18-5
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Color Colorless
Melting Point ∼0°C
Percent Purity 10%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (w/v) Aqueous Solution
Odor almond-like odor
Packaging Natural Poly Bottle
pH Alkaline
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Sodium Cyanide
Show More Show Less