missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyanate, 85%, pure

Catalog No. AC180562500
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC180562500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC180562500 Supplier Thermo Scientific Chemicals Supplier No. 180562500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 917-61-3 | CNNaO | 65.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 917-61-3
Molecular Formula CNNaO
Molecular Weight (g/mol) 65.01
MDL Number MFCD00011121
InChI Key ZVCDLGYNFYZZOK-UHFFFAOYSA-M
Synonym sodium cyanate, zassol, cyanic acid, sodium salt, sodiumcyanate, cyansan, weecon, sodium isocyanate, san-cyan, unii-8ufs3jrv8p, naocn
PubChem CID 517096
ChEBI CHEBI:38906
IUPAC Name sodium;cyanate
SMILES C(#N)[O-].[Na+]

Specifications

Melting Point 550°C
Color Beige or White to Yellow
Physical Form Crystalline Powder and/or Chunks
Quantity 250 g
Assay Percent Range 80% min. (Argentometry)
Linear Formula NaOCN
Beilstein 03, IV, 82
Merck Index 15, 8740
Solubility Information Solubility in water: 110g/L (20 c). Other solubilities: 0.22g/100g alcohol (0 c),insoluble in ether
Formula Weight 65.01
Percent Purity 85%
Grade Pure
Loss on Drying 3 % max. (5 g, 105°C, 2 hrs)
Packaging Plastic bottle
Chemical Name or Material Sodium cyanate, 85%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-030-6

RUO – Research Use Only