missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Citrate, For Copper, Certified, 30% (w/v), LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 74.84, 25.16 |
| Density | 1.2g/mL |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Specific Gravity | 1.2g/mL |
| Suitability | Passes Test |
| Decomposition Information | Fume; Carbon monoxide; Carbon dioxide |
| Linear Formula | Na3C6O7H5·2H2O |
| Quantity | 1 L |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |