accessibility menu, dialog, popup

UserName

CAS RN 63126-89-6

IUPAC Name: copper(2+) disodium 2-hydroxypropane-1,2,3-tricarboxylic acid hydrogen carbonate hydroxide sulfate
Synonyms: copper(2+) disodium citric acid bicarbonate hydroxide sulfate
Molecular Weight (g/mol): 475.73
Molecular Formula: C7H10CuNa2O15S
InChi Key: QFKWSRIUZIYLCK-UHFFFAOYSA-J
SMILES: [OH-].[Na+].[Na+].[Cu++].OC([O-])=O.[O-]S([O-])(=O)=O.OC(=O)CC(O)(CC(O)=O)C(O)=O
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results