missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride - Hydroxylamine Sulfate, For Metals, Certified, LabChem™
Specifications
Specifications
| Color | Colorless to Yellow |
| CAS Max % | 12, 76, 12 |
| Linear Formula | NaCl |
| Density | 1.1g/mL |
| Solubility Information | Soluble in water |
| Formula Weight | 58.44 |
| Grade | Certified |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Specific Gravity | 1.1g/mL |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. May cause an allergic skin reaction. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs (respiratory system, digestive system) through prolonged or repeated exposure (oral). Toxic to aquatic life. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Do not breathe mist. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. Contaminated work clothing must not be allowed out of the workplace. Avoid release to the environment. If exposed or concerned: Get medical advice/attention. If on skin: Wash with plenty of soap and water. Take off contaminated clothing and wash it before reuse. If skin irritation or rash occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |