missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride - Hydroxylamine Sulfate, For Metals, Certified, LabChem™

Catalog No. LC236001
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC236001 500 mL
LC236002 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC236001 Supplier LabChem Supplier No. LC236001
Only null left

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5 , 10039-54-0
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color Colorless to Yellow
CAS Max % 12, 76, 12
Linear Formula NaCl
Density 1.1g/mL
Solubility Information Soluble in water
Formula Weight 58.44
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.1g/mL
Suitability Passes Test
Decomposition Information Gaseous Ammonia; Hydrogen chloride
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Sodium Chloride - Hydroxylamine Sulfate, For Metals
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing cancer.
May cause damage to organs (respiratory system, digestive system) through prolonged or repeated exposure (oral).
Toxic to aquatic life.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature