missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, Crystal, ACS, 99%, Spectrum™ Chemical
$227.76 - $7,528.41
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| IUPAC Name | sodium chloride |
| SMILES | [Na+].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18613413
|
Spectrum Chemical Mfg Cor
S1240500GM |
500 g | Poly Bottle |
N/A
|
|
||||
|
18613419
|
Spectrum Chemical Mfg Cor
S1240350LBBL |
350 lb. | Fiber Drum |
N/A
|
|
||||
|
18613414
|
Spectrum Chemical Mfg Cor
S12401KG |
1 kg | Poly Bottle |
N/A
|
|
||||
|
18613415
|
Spectrum Chemical Mfg Cor
S124025KGGL |
2.5 kg | Amber Glass Bottle |
N/A
|
|
||||
|
18613416
|
Spectrum Chemical Mfg Cor
S124025KGPL |
2.5 kg | Poly Bottle |
N/A
|
|
||||
|
18613417
|
Spectrum Chemical Mfg Cor
S124012KG |
12 kg | Poly Pail |
N/A
|
|
||||
|
18613418
|
Spectrum Chemical Mfg Cor
S124045KGBL |
45 kg | Fiber Drum |
N/A
|
|
||||
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.