missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Crystal, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S124025KGGL

 View more versions of this product

Catalog No. 18613415


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7647-14-5
5.0 to 9.0
ClNa
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
99%
To Pass Test
1
2.5 kg
MFCD00003477
[Na+].[Cl-]
58.44
ACS
Amber Glass Bottle

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
[Na+].[Cl-]
ClNa
MFCD00003477
sodium chloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.