missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Crystal, ACS, 99%, Spectrum™ Chemical

Catalog No. 18613417
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
12 kg
45 kg
350 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613417 12 kg Poly Pail
18613413 500 g Poly Bottle
18613419 350 lb. Fiber Drum
18613414 1 kg Poly Bottle
18613415 2.5 kg Amber Glass Bottle
18613416 2.5 kg Poly Bottle
18613418 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18613417 Supplier Spectrum Chemical Mfg Cor Supplier No. S124012KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

pH 5.0 to 9.0
CAS Max % 1
Quantity 12 kg
Percent Purity 99%
Grade ACS
Identification To Pass Test
Packaging Poly Pail

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.