missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, ACS Grade, ≥ 99.0%, LabChem™
Specifications
Specifications
| Boiling Point | 1,413°C |
| Melting Point | 801°C |
| Color | White |
| CAS Max % | 100 |
| Physical Form | Crystals |
| Quantity | 500 g |
| Linear Formula | NaCl |
| Solubility Information | Soluble in water |
| Formula Weight | 58.44 |
| Percent Purity | ≥99.0% |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Product is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| EINECSNumber | 231-598-3 |
| Recommended Storage | Room Temperature |