missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride - Acetic Acid Standard, 2.00% (w/w) Sodium Chloride/1.00% (w/w) Acetic Acid, Ricca Chemical
Specifications
Specifications
| pH | Acidic |
| CAS Min % | 97%, 2%, 1% |
| CAS Max % | 97%, 2%, 1% |
| Appearance | Colorless Liquid |
| Quantity | 4 L |
| Packaging | Poly Bottle |
| Assay Percent Range | 1.999 to 2.001% |
| Solubility Information | Miscible in water |
| Percent Purity | 2%/1% w/w |
| Grade | Laboratory |
| Show More |
Safety and Handling
| ShelfLife | 12 Months |