missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, 5% (w/v), Ricca Chemical

Catalog No. 721516
Change view
Click to view available options
Quantity:
500 mL
1 L
55 gal.
Packaging:
Natural Poly Bottle
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
721516 500 mL Natural Poly Bottle
721532 1 L Natural Poly Bottle
721555 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 721516 Supplier Ricca Chemical Company Supplier No. 721516

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

CAS Min % 93.0, 4.67
CAS Max % 97.0, 4.86
Percent Purity 5%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 5% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
pH 6.7 to 7.3
Quantity 500 mL
Chemical Name or Material Sodium Chloride