missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, 30% (w/v), Ricca Chemical

Catalog No. p-2917048
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72204 120 mL
722016 500 mL
722032 1 L
72205 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 72204 Supplier Ricca Chemical Company Supplier No. 72204
Only null left

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

CAS Min % 75.0, 22.63
CAS Max % 78.0, 23.55
Percent Purity 30%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 30% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
pH 6.7 to 7.3
Quantity 120 mL
Chemical Name or Material Sodium Chloride