missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, 0.9% (w/v), Isotonic Saline, Ricca Chemical
Suitable for Use as Blood Bank Saline
$54.25 - $331.81
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
| PubChem CID | 5234 |
| ChEBI | CHEBI:26710 |
| IUPAC Name | sodium chloride |
| SMILES | [Na+].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
7210-16
|
Ricca Chemical Company
RI721016 |
500 mL | Natural Poly Bottle |
Each for $54.25
|
|
||||
|
7210-32
|
Ricca Chemical Company
721032 |
1 L | Natural Poly Bottle |
Each for $73.81
|
|
||||
|
7210-1
|
Ricca Chemical Company
72101 |
4 L | Natural Poly Bottle |
Each for $113.03
|
|
||||
|
7210-2-5
|
Ricca Chemical Company
721025 |
10 L | Cubitainer |
Each for $303.66
|
|
||||
|
7210-5
|
Ricca Chemical Company
72105 |
20 L | Cubitainer |
Each for $331.81
|
|
||||
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| sodium chloride |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
Specifications
| 7647-14-5,7732-18-5 | |
| MFCD00003477 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| 0.9% | |
| 0.9% (w/v) | |
| Aqueous | |
| 500 mL | |
| Sodium Chloride, Isotonic Saline |
| ClNa | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| Laboratory | |
| Natural Poly Bottle | |
| 6.7 to 7.3 | |
| Non-sterile |