missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, 0.9% (w/v), Isotonic Saline, Ricca Chemical
Suitable for Use as Blood Bank Saline
Supplier: Ricca Chemical Company 72105
Specifications
| Sodium Chloride | |
| 7647-14-5,7732-18-5 | |
| MFCD00003477 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| 0.9% | |
| 0.9% (w/v) | |
| Aqueous | |
| 20 L |
| Isotonic Saline | |
| ClNa | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| Laboratory | |
| Cubitainer | |
| 6.7 to 7.3 | |
| Non-sterile |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| sodium chloride |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| CHEBI:26710 | |
| [Na+].[Cl-] |