missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, 0.85% (w/v), N Saline, Ricca Chemical

Suitable for Use as Blood Bank Saline

$62.22 - $394.97

Chemical Identifiers

CAS 7732-18-5
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
7200-16
SDS View Documents
Ricca Chemical Company
720016
500 mL Natural Poly Bottle
Each for $62.22
Only null left
 
7200-32
View Documents
Ricca Chemical Company
720032
1 L Natural Poly Bottle
Each for $75.41
Only null left
 
7200-1
View Documents
Ricca Chemical Company
RI72001
4 L Natural Poly Bottle
Each for $120.09
Only null left
 
7200-2-5
View Documents
Ricca Chemical Company
720025
10 L Cubitainer
Each for $316.72
Only null left
 
7200-5
SDS View Documents
Ricca Chemical Company
72005
20 L Cubitainer
Each for $394.97
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5

Specifications

7732-18-5
Laboratory
Aqueous
500 mL
Sodium Chloride, 0.85% (w/v), Normal Saline
0.85%
Natural Poly Bottle
6.7 to 7.3
Non-sterile
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'