missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, 0.85% (w/v), N Saline, Ricca Chemical

Suitable for Use as Blood Bank Saline

Supplier:  Ricca Chemical Company 720025

 View more versions of this product

Catalog No. 7200-2-5


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Chloride, 0.85% (w/v)
7732-18-5
Laboratory
Aqueous
10 L
Normal Saline
0.85%
Cubitainer
6.7 to 7.3
Non-sterile

Chemical Identifiers

7732-18-5
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions