missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bromodifluoroacetate 98.0+%, TCI America™

Catalog No. S09085G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S09085G Supplier TCI America Supplier No. S09085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84349-27-9
Molecular Formula C2BrF2NaO2
Molecular Weight (g/mol) 196.911
MDL Number MFCD04038355
InChI Key CAQKQIYWKXZJGD-UHFFFAOYSA-M
Synonym Bromodifluoroacetic Acid Sodium Salt
PubChem CID 2782418
IUPAC Name sodium;2-bromo-2,2-difluoroacetate
SMILES C(=O)(C(F)(F)Br)[O-].[Na+]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 196.91
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Bromodifluoroacetate