missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bromate, Solstice

Catalog No. 6001377
Change view
Click to view available options
Quantity:
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001377 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001377 Supplier Solstice Supplier No. 0215125KG

≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7789-38-0
Molecular Formula BrNaO3
Molecular Weight (g/mol) 150.891
MDL Number MFCD00003476
InChI Key XUXNAKZDHHEHPC-UHFFFAOYSA-M
Synonym sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french
PubChem CID 23668195
ChEBI CHEBI:75229
IUPAC Name sodium;bromate
SMILES [O-]Br(=O)=O.[Na+]

Specifications

Quantity 25 kg
Linear Formula NaBrO3
UN Number 1494
Formula Weight 150.89g/mol
Percent Purity ≥99%
Density 3.339g/mL at 25°C
Chemical Name or Material Sodium Bromate
Hazard Statement P220-P261-P305 + P351 + P338
Precautionary Statement H272-H302-H315-H319-H335
EINECSNumber 232-160-4
RTECSNumber EF8750000