missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, (VenPureTM solution), 12% aq. solution, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 448510025

Numéro de catalogue. AC448510025

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Spécifications

Sodium borohydride
16940-66-2 , 1310-73-2 , 67-56-1 , 7732-18-5
BH4Na
MFCD00003518
SBH, Sodium tetrahydroborate
37.82
1.4
VenPure™ solution, 12% aq. solution
2.5L
NaBH4
15,8727
Solubility in water: completely soluble
37.82
1.4000g/mL
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May damage fertility.
May damage the unborn child.
May be corrosive to metals.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Rinse mouth.
Do NOT induce

GHS Signal Word: Danger

EINECSNumber : 241-004-4

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only