missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfite, Granular, ACS, 58.5%, Spectrum™ Chemical

Catalog No. 18613317
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613317 500 g Poly Pail
18613315 2.5 kg Poly Pail
18613314 12 kg Poly Pail
18613316 45 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613317 Supplier Spectrum Chemical Mfg Cor Supplier No. S1173500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7631-90-5

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 500 g
Molecular Weight (g/mol) 104.06
Percent Purity 58.5%
Grade ACS/Reagent
Identification To Pass Test
Packaging Poly Pail
Chemical Name or Material Sodium Bisulfite, Granular

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.