missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfite, ACS Reagent , Reagents

Catalog No. 01337757
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337757 12 kg
01337756 2.5 kg
01337755 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337757 Supplier Labchem Inc Supplier No. C131000012KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Bisulfite, ACS Reagent , ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7631-90-5
Molecular Formula NaO3S
Molecular Weight (g/mol) 103.05
InChI Key DWAQJAXMDSEUJJ-UHFFFAOYSA-L
Synonym Sodium Hydrogen Sulfite
IUPAC Name sodium sulfite
SMILES [Na+].[O-]S([O-])=O

Specifications

Boiling Point 146°C
Melting Point 5°C
Color Colorless/White
Physical Form Solid
Quantity 12 kg
Grade ACS
Packaging Poly Pail
Specific Gravity 1.48
Chemical Name or Material Sodium Bisulfite, ACS Reagent *This reagent is usually a mixture containing some Sodium Metabisulfite (CAS# 7681-57-4).