missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfite, 35% w/w, Ricca Chemical Company

Catalog No. 7149600120A
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7149600120A 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 7149600120A Supplier Ricca Chemical Company Supplier No. R7149600120A*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analytical reagent grade solution of sodium bisulfite.

Specifications

Color Colorless to Yellow
CAS 7631-90-5, 7732-18-5
Grade Laboratory
Concentration or Composition (by Analyte or Components) 35T w/w
Specific Gravity ~1.37
Quantity 120 mL
Physical Form Liquid
Chemical Name or Material Sodium Bisulfite Solution
ShelfLife 12 Months