missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfite, 10% (w/v), Ricca Chemical

Catalog No. 714932
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
714932 1 L Natural Poly Bottle
714916 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 714932 Supplier Ricca Chemical Company Supplier No. 714932

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

APHA for Manganese Testing

Chemical Identifiers

CAS 7631-90-5 , 7732-18-5
Molecular Formula NaO3S
Molecular Weight (g/mol) 103.05
MDL Number MFCD00003530
InChI Key DWAQJAXMDSEUJJ-UHFFFAOYSA-L
Synonym sodium bisulfite, sodium hydrogen sulfite, sodium bisulphite, sodium hydrogensulfite, sodium sulfhydrate, monosodium sulfite, uantax sbs, sulfurous acid, monosodium salt, caswell no. 750, hydrogen sodium sulfite
PubChem CID 23665763
ChEBI CHEBI:26709
IUPAC Name sodium sulfite
SMILES [Na+].[O-]S([O-])=O

Specifications

Color Colorless
CAS Min % 89.0, 8.91
CAS Max % 93.0, 9.28
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH Acidic
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 10% (w/v)
Chemical Name or Material Sodium Bisulfite
Show More Show Less