missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bis(trimethylsilyl)amide (contains 2-Methyl-2-butene) (38% in Tetrahydrofuran, ca. 1.9mol/L), TCI America™

Catalog No. H0894100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0894100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H0894100ML Supplier TCI America Supplier No. H0894100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1070-89-9
Molecular Formula C6H18NNaSi2
Molecular Weight (g/mol) 183.377
MDL Number MFCD00009835
InChI Key WRIKHQLVHPKCJU-UHFFFAOYSA-N
Synonym sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt
PubChem CID 2724254
IUPAC Name sodium;bis(trimethylsilyl)azanide
SMILES C[Si](C)(C)[N-][Si](C)(C)C.[Na+]

Specifications

Quantity 100 mL
UN Number 2924
Formula Weight 183.38
Physical Form Liquid
Chemical Name or Material Sodium Bis(trimethylsilyl)amide (contains 2-Methyl-2-butene) (38% in Tetrahydrofuran, ca. 1.9mol/L)