missing translation for 'onlineSavingsMsg'
Learn More

Sodium bis(trimethylsilyl)amide, 2M soln. in THF

Catalog No. AAL13352AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13352AP 500 mL
AAL13352AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL13352AP Supplier Thermo Scientific Chemicals Supplier No. L13352AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1070-89-9 | C6H18NNaSi2 | 183.377 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bis(trimethylsilyl)amide is a strong base used in organic synthesis for deprotonation reactions and base catalyzed reaction. It is also used to enhance the polymerization of pheneylacetylene in conjunction with rhodium(I) catalysis. It is involved in the generation of enolates, Wittig reagents and carbenes.

Solubility
Miscible with benzene and toluene.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, strong oxidizing agents and oxygen. Dry residue is explosive.

Chemical Identifiers

CAS 1070-89-9
Molecular Formula C6H18NNaSi2
Molecular Weight (g/mol) 183.377
MDL Number MFCD00009835
InChI Key WRIKHQLVHPKCJU-UHFFFAOYSA-N
Synonym sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt
PubChem CID 2724254
IUPAC Name sodium;bis(trimethylsilyl)azanide
SMILES C[Si](C)(C)[N-][Si](C)(C)C.[Na+]

Specifications

Quantity 500 mL
Flash Point −17°C (1°F)
Sensitivity Moisture sensitive; May form a precipitate
Density 0.92
UN Number UN2924
Beilstein 3629917
Solubility Information Miscible with benzene and toluene.
Formula Weight 183.38
Concentration or Composition (by Analyte or Components) 2M soln. in THF
Chemical Name or Material Sodium bis(trimethylsilyl)amide
Hazard Category H225-H302-H314-H335-H336-H341-H351
Hazard Statement GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 213-983-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only